Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558321 Supplier Invitrogen™ Supplier No. PA558321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGFSDIHFNS RNTFQVSRGQ SARDHLPPAG PPVPLPAAEQ GPAGASARAR RSGGFADFFT RNLFPKRTKE LKSVVHSAPG WKLFGKVPPR ENLQKTSKII QQEYEART Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D12
Gene Accession No. O60347
Gene Alias KIAA0608; TBC1 domain family member 12; TBC1 domain family, member 12; TBC1D12
Gene Symbols TBC1D12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D12. Recombinant protein control fragment (Product #RP-97059).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.