Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D13 Antibody (OTI2C5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245985
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
TBC1D13 Monoclonal specifically detects TBC1D13 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TBC1D13 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
FLJ10743, TBC1 domain family member 13, TBC1 domain family, member 13 | |
Mouse | |
46.4 kDa | |
0.1 mL | |
Signal Transduction | |
54662 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2C5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
TBC1D13 | |
Full length human recombinant protein of human TBC1D13 (NP_060671) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction