Learn More
Invitrogen™ TBC1D13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114139
Description
Immunogen sequence: FLLLVCCAML MLIREQLLEG DFTVNMRLLQ DYPITDVCQI LQKAKELQDS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
TBC1D13 may act as a GTPase-activating protein for Rab family protein(s).
Specifications
TBC1D13 | |
Polyclonal | |
Unconjugated | |
TBC1D13 | |
2600014A06Rik; BC025586; TBC1 domain family member 13; TBC1 domain family, member 13; TBC1D13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54662 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NVG8 | |
TBC1D13 | |
Synthetic peptide directed towards the middle region of human TBC1D13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.