Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D14 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306257PCP
Description
TBC1D14 Polyclonal antibody specifically detects TBC1D14 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TBC1D14 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
KIAA1322FLJ32400, TBC1 domain family member 14, TBC1 domain family, member 14 | |
Produced in rabbits immunized with E. coli-derived Human TBC1D14 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57533 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction