Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220577
Description
TBC1D15 Polyclonal specifically detects TBC1D15 in Human samples. It is validated for Western Blot.Specifications
TBC1D15 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
domain family member 15, TBC1 domain family, member 15 | |
Rabbit | |
79 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (88%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q8TC07 | |
TBC1D15 | |
Recombinant protein encompassing a sequence within the N-terminus region of human TBC1D15. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
64786 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction