Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553053 Supplier Invitrogen™ Supplier No. PA553053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSKLLIESLE KYVVLCESPQ DKRTLLVNCQ NKSLSQSFEN LLDEPAYGLI QKIKKDPYTA TMIGFSKVTN YIFDSLRGSD PSTHQRPPSE MADFLSDAIP GLKINQQEEP GFEVITRI Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 84%.

Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D15
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D15
Gene Accession No. Q8TC07, Q9CXF4
Gene Alias 4432405K22Rik; AU022358; AU023730; AW557774; GAP for RAB7; GTPase-activating protein RAB7; Ly6dl; lymphocyte antigen 6, locus D ligand; Rab7-GAP; TBC1 domain family member 15; TBC1 domain family, member 15; Tbc1d15; Tre2/Bub2/Cdc16 domain family member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15
Gene Symbols TBC1D15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D15. Recombinant protein control fragment (Product #RP-90741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366896, 64786, 66687
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.