Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598645
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598645 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598645 Supplier Invitrogen™ Supplier No. PA598645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D15 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TBC1D15
Gene Accession No. Q8TC07
Gene Alias 4432405K22Rik; AU022358; AU023730; AW557774; GAP for RAB7; GTPase-activating protein RAB7; Ly6dl; lymphocyte antigen 6, locus D ligand; Rab7-GAP; TBC1 domain family member 15; TBC1 domain family, member 15; Tbc1d15; Tre2/Bub2/Cdc16 domain family member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15
Gene Symbols TBC1D15
Host Species Rabbit
Immunogen Recombinant Human TBC1 domain family member 15 protein (283-404AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64786
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.