Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555089
Description
Immunogen sequence: FSSPFCLSPI SAALAESRGS VFLESDSSPP SSSDAGLRFP DSNGLLQTPR WDEPQRVCAL EQICGVFRVD LGHMRSLRLF FSDEACTSGQ LVVASRESQY KVFHFHHGGL DKLSDVFQQW KYCTEMQLKD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
May act as a GTPase-activating protein for Rab family protein(s).
Specifications
TBC1D16 | |
Polyclonal | |
Unconjugated | |
TBC1D16 | |
CTD-2529O21.1; TBC1 domain family member 16; TBC1 domain family, member 16; TBC1D16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
125058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TBP0 | |
TBC1D16 | |
Recombinant protein corresponding to Human TBC1D16. Recombinant protein control fragment (Product #RP-93369). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction