Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566800 Supplier Invitrogen™ Supplier No. PA566800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FFGGIDVSIRG EVWPFLLRYY SHESTSEERE ALRLQKRKEY SEIQQKRLSM TPEEHRAFWR NVQFTVDKDV VRTDRNNQFF RGEDNPNVES MRRILLNYAV YNPAV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

May act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D16
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D16
Gene Accession No. Q8TBP0
Gene Alias CTD-2529O21.1; TBC1 domain family member 16; TBC1 domain family, member 16; TBC1D16
Gene Symbols TBC1D16
Host Species Rabbit
Immunogen Recombinant Human TBC1D16. Recombinant protein control fragment (Product #RP-105372).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.