Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570937 Supplier Invitrogen™ Supplier No. PA570937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Goat: 80%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%; Zebrafish: 77%.

Probable GTPase-activating protein for Rab8; its transient association with Rab8 is mediated by OPTN. Inhibits Rab8-mediated endocytic trafficking, such as of transferrin receptor (TfR) and reduces Rab8 recruitnment to tubules emanating from the endocytic recycling compartment (ERC). Involved in regulation of autophagy. Mediates inhibition of autophagy caused by the OPTN variant GLC1E LYS-50; the function requires its catalytic activity, however, the involved Rab is not known. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TBC1D17
Gene Accession No. Q8BYH7
Gene Alias BC017607; hypothetical protein LOC554092; TBC1 domain family member 17; TBC1 domain family, member 17; TBC1D17; wu:fc56c02; zgc:110443; zgc:110443 protein
Gene Symbols TBC1D17
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse TBC1D17.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233204
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.