Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556577 Supplier Invitrogen™ Supplier No. PA556577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLMDKNHAKQ QVICKLSEKV TQDFTHPPDQ SPLRPDAANR DFLSQQGKIE HLKDDMEAYR TQNCFLNSEI HQVTKIWRKV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Acts as GTPase-activating protein for RAB7A. Signal effector acting as a linker between RAC1 and RAB7A, leading to RAB7A inactivation and subsequent inhibition of cadherin degradation and reduced cell-cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D2
Gene Accession No. Q9BYX2
Gene Alias A630005A06Rik; armus; BC022630; BC1D2; Gm1037; likely ortholog of H. sapiens TBC1 domain family, member 2 (TBC1D2); PARIS1; PARIS-1; PP8997; Prostate antigen recognized and identified by SEREX 1; TBC1 domain family member 2; TBC1 domain family member 2A; TBC1 domain family, member 2; TBC1 domain family, member 2A; TBC1D2; TBC1D2A
Gene Symbols TBC1D2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D2. Recombinant protein control fragment (Product #RP-92894).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55357
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.