Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104176 Supplier Invitrogen™ Supplier No. PA5104176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBC1D2.

Acts as GTPase-activating protein for RAB7A. Signal effector acting as a linker between RAC1 and RAB7A, leading to RAB7A inactivation and subsequent inhibition of cadherin degradation and reduced cell-cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBC1D2
Gene Accession No. B5DFA1, Q9BYX2
Gene Alias A630005A06Rik; armus; BC022630; BC1D2; Gm1037; likely ortholog of H. sapiens TBC1 domain family, member 2 (TBC1D2); PARIS1; PARIS-1; PP8997; Prostate antigen recognized and identified by SEREX 1; TBC1 domain family member 2; TBC1 domain family member 2A; TBC1 domain family, member 2; TBC1 domain family, member 2A; TBC1D2; TBC1D2A
Gene Symbols TBC1D2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBC1D2(Accession Q9BYX2), corresponding to amino acid residues R62-E112.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313234, 55357
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.