Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140188
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VAEKEKALLT KCAYLQARNC QVESKYLAGL RRLQEALGDE ASECSELLRQ LVQEALQWEA GEASSDSIEL SPISKYDEYG FLTVPDYEVE DLKLL.
Acts as GTPase-activating protein for RAB7A. Signal effector acting as a linker between RAC1 and RAB7A, leading to RAB7A inactivation and subsequent inhibition of cadherin degradation and reduced cell-cell adhesion.
Specifications
TBC1D2 | |
Polyclonal | |
Unconjugated | |
TBC1D2 | |
A630005A06Rik; armus; BC022630; BC1D2; Gm1037; likely ortholog of H. sapiens TBC1 domain family, member 2 (TBC1D2); PARIS1; PARIS-1; PP8997; Prostate antigen recognized and identified by SEREX 1; TBC1 domain family member 2; TBC1 domain family member 2A; TBC1 domain family, member 2; TBC1 domain family, member 2A; TBC1D2; TBC1D2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYX2 | |
TBC1D2 | |
Recombinant protein corresponding to Human TBC1D2. Recombinant protein control fragment (Product #RP-109561). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction