Learn More
TBC1D2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303441AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Acts as GTPase-activating protein for RAB7A. Signal effector acting as a linker between RAC1 and RAB7A, leading to RAB7A inactivation and subsequent inhibition of cadherin degradation and reduced cell-cell adhesion.Specifications
TBC1D2 | |
Polyclonal | |
Unconjugated | |
TBC1D2 | |
TBC1 domain family member 2A, Armus, Prostate antigen recognized and identified by SEREX 1, PARIS-1, TBC1D2, PARIS1, PP8997, TBC1D2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55357 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9BYX2 | |
TBC1D2 | |
Between 225 and 275 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.