Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113235
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113235 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113235 Supplier Invitrogen™ Supplier No. PA5113235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D20 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

GTPase-activating protein specific for Rab1 and Rab2 small GTPase families for which it can accelerate the intrinsic GTP hydrolysis rate by more than five orders of magnitude.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D20
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TBC1D20
Gene Accession No. Q96BZ9, Q9D9I4
Gene Alias 1110028I04Rik; 2810442O16Rik; AI414693; C20orf140; TBC1 domain family member 20; TBC1 domain family, member 20; TBC1D20; WARBM4
Gene Symbols TBC1D20
Host Species Rabbit
Immunogen Recombinant Human TBC1 domain family member 20 protein (1-237AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128637, 67231
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.