Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114201 Supplier Invitrogen™ Supplier No. PA5114201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERTAASTFKD FELASAQQRP DMVLRQRFRG LLRPEDRTKD VLTKPRTNRF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%.

GTPase-activating protein specific for Rab1 and Rab2 small GTPase families for which it can accelerate the intrinsic GTP hydrolysis rate by more than five orders of magnitude.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D20
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TBC1D20
Gene Accession No. Q96BZ9
Gene Alias 1110028I04Rik; 2810442O16Rik; AI414693; C20orf140; TBC1 domain family member 20; TBC1 domain family, member 20; TBC1D20; WARBM4
Gene Symbols TBC1D20
Host Species Rabbit
Immunogen Immunogen sequence: ERTAASTFKD FELASAQQRP DMVLRQRFRG LLRPEDRTKD VLTKPRTNRF.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128637
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.