Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D21 Antibody (OTI2A6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274451MFV450
Description
TBC1D21 Monoclonal antibody specifically detects TBC1D21 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
TBC1D21 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2A6 | |
Flow Cytometry | |
MGC34741, TBC1 domain family member 21, TBC1 domain family, member 21 | |
Full length human recombinant protein of human TBC1D21(NP_699187) produced in HEK293T cell. | |
0.1 mL | |
GPCR, Signal Transduction | |
161514 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction