Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D21 Antibody (OTI2A6), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274451PECY55
Description
TBC1D21 Monoclonal specifically detects TBC1D21 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
TBC1D21 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
TBC1D21 | |
Full length recombinant protein of human TBC1D21 (NP_699187) produced in HEK293T cells. | |
0.1 mL | |
161514.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI2A6 | |
Flow Cytometry | |
MGC34741, TBC1 domain family member 21, TBC1 domain family, member 21 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction