Learn More
Invitrogen™ TBC1D21 Monoclonal Antibody (OTI1C7)

Description
TBC1D21 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TBC1D21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C7 |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TBC1D21 |
| Gene Accession No. | Q8IYX1 |
| Gene Alias | male germ cell-specific expressed, containing a RabGAP domain; MgcRabGAP; TBC1 domain family member 21; TBC1 domain family, member 21; TBC1D21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.