Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560321 Supplier Invitrogen™ Supplier No. PA560321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLSPENSLSA RQSASFILVK RKPPIDKTEW DSFFDESGHL AKSRDFICVN ILERGLHPFV RTEAWKFLTG YFSWQSSQDE R Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.

TBC1D21 may act as a GTPase-activating protein for Rab family protein(s). May be involved in acrosome formation and cytoskeletal reorganization during spermiogenesis, possibly by regulating RAB3A activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D21
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D21
Gene Accession No. Q8IYX1
Gene Alias male germ cell-specific expressed, containing a RabGAP domain; MgcRabGAP; TBC1 domain family member 21; TBC1 domain family, member 21; TBC1D21
Gene Symbols TBC1D21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D21. Recombinant protein control fragment (Product #RP-98164).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161514
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.