Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D22A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186811
Description
TBC1D22A Polyclonal specifically detects TBC1D22A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TBC1D22A | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
HSC79E021, TBC1 domain family member 22A, TBC1 domain family, member 22A | |
Rabbit | |
Affinity Purified | |
RUO | |
25771 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TBC1D22A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:HPASGYVQGINDLVTPFFVVFICEYIEAEEVDTVDVSGVPAEVLCNIEADTYWCMSKLLDGIQDNYTFAQPGIQMKVKMLEELVSRIDEQVHRHLDQHEVRYLQFAFRWMNNLLMREVPLRCTIRLWDTYQSE | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction