Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D22A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551526 Supplier Invitrogen™ Supplier No. PA551526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPASGYVQGI NDLVTPFFVV FICEYIEAEE VDTVDVSGVP AEVLCNIEAD TYWCMSKLLD GIQDNYTFAQ PGIQMKVKML EELVSRIDEQ VHRHLDQHEV RYLQFAFRWM NNLLMREVPL RCTIRLWDTY QSE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 59%.

TBC1D22A (TBC1 domain family, member 22A), also known as C22orf4, is a 517 amino acid protein that contains one Rab-GAP TBC domain and is thought to function as a GTPase-activating protein for Rab family members. Multiple isoforms of TBC1D22A exist due to alternative splicing events. The gene encoding TBC1D22A maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia. Additionally, translocations between chromosomes 9 and 22 may lead to the formation of the Philadelphia chromosome and the subsequent production of the novel fusion protein BCR-Abl, a potent cell proliferation activator found in several types of leukemias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D22A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D22A
Gene Accession No. Q8WUA7
Gene Alias BC023106; C22orf4; D15Ertd781e; HSC79E021; putative GTPase activator; RGD1306588; TBC1 domain family member 22A; TBC1 domain family, member 22a; TBC1D22A
Gene Symbols TBC1D22A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D22A. Recombinant protein control fragment (Product #RP-91366).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.