Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D22B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555727
Description
Immunogen sequence: QHPPLDPRLT KNFIKERSKV NTVPLKNKKA SSFHEFARNT SDAWDIGDDE EEDFSSPSFQ TLNSK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

Specifications
TBC1D22B | |
Polyclonal | |
Unconjugated | |
TBC1D22B | |
C6orf197; TBC1 domain family member 22B; TBC1 domain family, member 22B; TBC1D22B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NU19 | |
TBC1D22B | |
Recombinant protein corresponding to Human TBC1D22B. Recombinant protein control fragment (Product #RP-94625). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction