Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558274 Supplier Invitrogen™ Supplier No. PA558274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLSTAFHLDS DLMLQNPSEF AQSVKSLLEA QKQSIESGSI AGGEHLCFMG SGREEEDMYM NMVLAHFLQK NKEYVSIASG GFMALQQHLA DINVDGPENG Y Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Putative Rab GTPase-activating protein which plays a role in vesicular trafficking (PubMed:28823707). Involved in endosome-to-Golgi trafficking. Acts as a bridging protein by binding simultaneously to golgins, including GOLGA1 and GOLGA4, located at the trans-Golgi, and to the WASH complex, located on endosome-derived vesicles (PubMed:29084197, PubMed:29426865). Together with WDR11 complex facilitates the golgin-mediated capture of vesicles generated using AP-1 (PubMed:29426865). Plays a role in brain development, including in cortical neuron positioning. May also be important for neurite outgrowth, possibly through its involvement in membrane trafficking and cargo delivery, 2 processes that are essential for axonal and dendritic growth. May act as a general inhibitor of innate immunity signaling, strongly inhibiting multiple TLR and dectin/CLEC7A-signaling pathways. Does not alter initial activation events, but instead affects maintenance of inflammatory gene expression several hours after bacterial lipopolysaccharide (LPS) challenge. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D23
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D23
Gene Accession No. Q9NUY8
Gene Alias 4930451A13Rik; AU015720; AU043671; AU043778; D030022P07Rik; HCV non-structural protein 4A-transactivated protein 1; HCV nonstructural protein 4A-transactivated protein 1; hypothetical protein LOC393459; NS4ATP1; RGD1307925; TBC1 domain family member 23; TBC1 domain family, member 23; TBC1D23; W51689; zgc:63756
Gene Symbols TBC1D23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D23. Recombinant protein control fragment (Product #RP-96258).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.