Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560800 Supplier Invitrogen™ Supplier No. PA560800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGKVYQRLIR DIPCRTVTPD ASVYSDIVGK IVGKHSSSCL PLPEFVDNTQ VPSYCLNARG EGAVRKILLC LANQFPD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

TBC1D24 may act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D24
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D24
Gene Accession No. Q9ULP9
Gene Alias 9630033P11; C530046L02Rik; DFNA65; DFNB86; DOORS; EIEE16; FIME; KIAA1171; MGC137396 protein; mKIAA1171; RGD1306143; skywalker homolog; TBC/LysM-associated domain containing 6; TBC1 domain family member 24; TBC1 domain family, member 24; Tbc1d24; TLDC6
Gene Symbols TBC1D24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D24. Recombinant protein control fragment (Product #RP-98706).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.