Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556082 Supplier Invitrogen™ Supplier No. PA556082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVDPQITSLD VLQHILIRAF DLSGKKNFGI SYLGRDRLGQ EVYLSLLSDW DLSTAFATAS KPYLQLRVDI RPSEDSPLLE DWDIISPKDV SGSDVLLAEK RSSLTTAALP FTQSIL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

TBC1D25 encodes a protein with a TBC domain and functions as a Rab GTPase activating protein. The encoded protein is involved in the fusion of autophagosomes with endosomes and lysosomes. TBC1D25 was previously known as ornithine aminotransferase-like 1, but has no similarity to ornithine aminotransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D25
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D25
Gene Accession No. Q3MII6
Gene Alias 5SN3 snoRNA; 6330576A08Rik; A530047E07; DXHXS7927e; MG81; mMg81; OATL1; ornithine aminotransferase-like 1; TBC1 domain family member 25; TBC1 domain family, member 25; Tbc1d25
Gene Symbols TBC1D25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D25. Recombinant protein control fragment (Product #RP-91987).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.