Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562193 Supplier Invitrogen™ Supplier No. PA562193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAGERHSLWY STAQILPGSR GSYRTRSRCC TSPSQRS Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 38%.

TBC1D26 may act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D26
Gene Accession No. Q86UD7
Gene Alias TBC1 domain family member 26; TBC1 domain family, member 26; TBC1D26
Gene Symbols TBC1D26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D26. Recombinant protein control fragment (Product #RP-100341).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.