Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D28 Antibody (OTI4A9), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274461IR
Description
TBC1D28 Monoclonal specifically detects TBC1D28 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TBC1D28 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TBC1D28 | |
Full length recombinant protein of human TBC1D28 (NP_001034486) produced in E.coli. | |
0.1 mL | |
254272.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4A9 | |
Western Blot, Immunohistochemistry | |
FLJ40244, MGC138217, MGC138221, TBC1 domain family member 28, TBC1 domain family, member 28 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction