Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D28 Monoclonal Antibody (OTI4A9)
GREENER_CHOICE

Catalog No. PIMA526953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526953 Supplier Invitrogen™ Supplier No. MA526953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBC1D28 Monoclonal Antibody for Western Blot, IHC (P)

TBC1D28 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D28
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TBC1D28
Gene Accession No. Q2M2D7
Gene Alias TBC1 domain family member 28; TBC1 domain family, member 28; TBC1D28
Gene Symbols TBC1D28
Host Species Mouse
Immunogen Full length human recombinant protein of TBC1D28 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254272
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.