Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562662 Supplier Invitrogen™ Supplier No. PA562662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSSQYDKYF TSSRLCGGVP KDTLELLHQK DDQILGLTSQ LERFSLEKES LQQEVRTLKS KVGELNEQLG MLMETIQAKD E Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

TBC1D2B is a protein coding gene. May act as a GTPase-activating protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D2B
Gene Accession No. Q9UPU7
Gene Alias KIAA1055; TBC1 domain family member 2B; TBC1 domain family, member 2B; TBC1D2B
Gene Symbols TBC1D2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D2B. Recombinant protein control fragment (Product #RP-98137).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23102
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.