Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114250 Supplier Invitrogen™ Supplier No. PA5114250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRFVDTWARD EDTVLKHLRA SMKKLTRKQG DLPPPAKPEQ GSSASRPVPA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

Acts as a GTPase activating protein for RAB5. Does not act on RAB4 or RAB11. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TBC1D3
Gene Accession No. Q8IZP1
Gene Alias PRC17; Prostate cancer gene 17 protein; protein TRE17-alpha; Rab GTPase-activating protein PRC17; TBC1 domain family member 3; TBC1 domain family member 3C; TBC1 domain family member 3F; TBC1 domain family, member 3; TBC1 domain family, member 3F; TBC1D3; TBC1D3A; TBC1D3C; TBC1D3D; TBC1D3E; TBC1D3F
Gene Symbols TBC1D3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human TBC1D3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729873
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.