Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558923
Description
Immunogen sequence: NSSPVINHLL LGKKMKMTNR AAKNAVIHIP GHTGGKISPV PYEDLKTKLN SPWRTHIRVH KKNMPRTKSH PGCGDTVGLI DEQNEASKTN GLG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

Specifications
TBC1D30 | |
Polyclonal | |
Unconjugated | |
TBC1D30 | |
KIAA0984; TBC1 domain family member 30; TBC1 domain family, member 30; TBC1D30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23329 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2I9 | |
TBC1D30 | |
Recombinant protein corresponding to Human TBC1D30. Recombinant protein control fragment (Product #RP-97694). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction