Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561582 Supplier Invitrogen™ Supplier No. PA561582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWQAEHPAER VRSAFAAPST DSDQGT Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

TBC1D3G acts as a GTPase activating protein for RAB5. Does not act on RAB4 or RAB11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D3C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D3C
Gene Accession No. Q6IPX1
Gene Alias PRC17; Prostate cancer gene 17 protein; Protein TRE17-alpha; Rab GTPase-activating protein PRC17; TBC1 domain family member 3; TBC1 domain family member 3C; TBC1 domain family member 3C/3D; TBC1 domain family member 3C-like protein ENSP00000341742; TBC1 domain family member 3G; TBC1 domain family member-like; TBC1 domain family, member 3C; TBC1 domain family, member 3G; TBC1D3; TBC1D3A; TBC1D3C; TBC1D3D; TBC1D3G
Gene Symbols TBC1D3C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D3C. Recombinant protein control fragment (Product #RP-104429).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.