Learn More
Invitrogen™ TBC1D3C Polyclonal Antibody

Description
This target displays homology in the following species: Human: 100%.
Specifications
Specifications
| Antigen | TBC1D3C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TBC1D3C |
| Gene Accession No. | Q6IPX1 |
| Gene Alias | PRC17; Prostate cancer gene 17 protein; Protein TRE17-alpha; Rab GTPase-activating protein PRC17; TBC1 domain family member 3; TBC1 domain family member 3C; TBC1 domain family member 3C/3D; TBC1 domain family member 3C-like protein ENSP00000341742; TBC1 domain family member 3G; TBC1 domain family member-like; TBC1 domain family, member 3C; TBC1 domain family, member 3G; TBC1D3; TBC1D3A; TBC1D3C; TBC1D3D; TBC1D3G |
| Gene Symbols | TBC1D3C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.