Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D3F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548350
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548350 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548350 Supplier Invitrogen™ Supplier No. PA548350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D3F Polyclonal Antibody for Western Blot

Acts as a GTPase activating protein for RAB5. Does not act on RAB4 or RAB11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D3F
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TBC1D3F
Gene Accession No. A6NER0
Gene Alias prostate cancer gene 17 protein; protein TRE17-alpha; TBC1 domain family member 3; TBC1 domain family member 3F; TBC1 domain family, member 3F; TBC1D3; TBC1D3E; TBC1D3F
Gene Symbols TBC1D3F
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 498-527 amino acids from the C-terminal region of human TBC1D3E.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.