Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D4 Antibody (OTI5E6), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274462PECY7
Description
TBC1D4 Monoclonal specifically detects TBC1D4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
TBC1D4 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
TBC1D4 | |
Full length recombinant protein of human TBC1D4 (NP_0055647) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Flow Cytometry | |
OTI5E6 | |
Flow Cytometry | |
Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
Mouse | |
Affinity Purified | |
Diabetes Research, Lipid and Metabolism, Phospho Specific | |
9882.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction