Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D4 Monoclonal Antibody (OTI5A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502707S


Description
TBC1D4 Monoclonal antibody specifically detects TBC1D4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
TBC1D4 | |
Monoclonal | |
0.41 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P08559 | |
Mouse | |
Affinity chromatography | |
RUO | |
5160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI5A6 | |
Unconjugated | |
PDHA1 | |
5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4 | |
Full length human recombinant protein of human PDHA1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction