Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D4, p Thr642 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244201 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244201 Supplier Novus Biologicals Supplier No. NBP244201
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TBC1D4 in Human, Mouse, Rat samples, and it is validated for ELISA, Western Blotting

TBC1D4 Polyclonal specifically detects TBC1D4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen TBC1D4 (p Thr642)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1ug/ml, ELISA 1:10000, Immunohistochemistry
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with No Preservative
Gene Accession No. Q8BYJ6
Gene Alias Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4
Gene Symbols TBC1D4
Host Species Rabbit
Immunogen TBC1D4 [p Thr642] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide surrounding the pT649 site of human TBC1D4 [p Thr642]. (Uniprot: Q8BYJ6)
Molecular Weight of Antigen 147.45 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9882
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.