Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D4, p Thr642 Antibody (SR1272), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32260750UL
Description
TBC1D4 Monoclonal antibody specifically detects TBC1D4 in Human samples. It is validated for Western BlotSpecifications
TBC1D4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1272 | |
Western Blot 1:500-1:2000 | |
Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
A synthesized peptide derived from human phospho-TBC1D4 [Thr642] (Uniprot #: O60343) | |
50 μg | |
Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
9882 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction