Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D5 Recombinant Rabbit Monoclonal Antibody (HL2390)
GREENER_CHOICE

Catalog No. FEMA550515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550515 Supplier Invitrogen™ Supplier No. MA550515
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

TBC1D5 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

TBC1 domain family member 5 (TBC1D5) may act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2390
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TBC1D5
Gene Accession No. Q92609
Gene Alias 1600014N05Rik; KIAA0210; RGD1561626; TBC1 domain family member 5; TBC1 domain family, member 5; TBC1D5
Gene Symbols TBC1D5
Host Species Rabbit
Immunogen Synthetic peptide of human TBC1D5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9779
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.