Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571263
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 93%.
This gene encodes a protein with a TBC (Tre-2/Bub2/CDC16) domain. Some mammalian proteins with this domain have been shown to function as Rab-GAPs by binding to specific Rab proteins and affecting their GTPase activity. Alternative splicing results in multiple transcript variants.
Specifications
TBC1D8B | |
Polyclonal | |
Unconjugated | |
TBC1D8B | |
4921505D17Rik; 9030605E16Rik; GRAMD8B; RGD1563084; RP11-321G1.1; TBC1 domain family member 8B; TBC1 domain family, member 8B; TBC1 domain family, member 8B (with GRAM domain); TBC1D8B | |
Rabbit | |
Affinity chromatography | |
RUO | |
54885 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q0IIM8 | |
TBC1D8B | |
synthetic peptide directed towards the N-terminal region of human TBC1D8B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction