Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D9 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055321DL594
Description
TBC1D9 Polyclonal antibody specifically detects TBC1D9 in Human samples. It is validated for Western BlotSpecifications
TBC1D9 | |
Polyclonal | |
Western Blot | |
KIAA0882TBC1 domain family member 9, MDR1, TBC1 domain family member 9A, TBC1 domain family, member 9, TBC1 domain family, member 9 (with GRAM domain), TBC1D9A | |
The immunogen recognized by this antibody maps to a region between residue 1118 and the C-terminus (residue 1168) of human TBC1 domain family, member 9 using the numbering given in entry NP_055945.1 (GeneID 23158). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23158 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction