Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055321
Description
TBC1D9 Polyclonal specifically detects TBC1D9 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
TBC1D9 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
KIAA0882TBC1 domain family member 9, MDR1, TBC1 domain family member 9A, TBC1 domain family, member 9, TBC1 domain family, member 9 (with GRAM domain), TBC1D9A | |
Rabbit | |
143 kDa | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque,European domestic ferret, African elephant, Gray short-tailed opossum and Northern white-cheekedgibbon. | |
Store at 4C. Do not freeze. |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 2-5 ug/mg lysate | |
Q6ZT07 | |
TBC1D9 | |
The immunogen recognized by this antibody maps to a region between residue 1118 and the C-terminus (residue 1168) of human TBC1 domain family, member 9 using the numbering given in entry NP_055945.1 (GeneID 23158). | |
Affinity Purified | |
RUO | |
23158 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction