Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552305 Supplier Invitrogen™ Supplier No. PA552305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGKLFVAQPA KEGGSGGSGP SCHQGIPGVL FPKKGPGQPY VVESVEPLPA SLAPDSEEHS LGGQMEDIKL EDSSPRDNGA CSSMLISDDD TKDDSSMSSY SVLSAGSHEE DKLHCEDI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

TBC1D9 may act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D9
Gene Accession No. Q6ZT07
Gene Alias GRAMD9; KIAA0882; MDR1; TBC1 domain family member 9; TBC1 domain family member 9A; TBC1 domain family, member 9 (with GRAM domain); TBC1D9; TBC1D9A
Gene Symbols TBC1D9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D9. Recombinant protein control fragment (Product #RP-93714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23158
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.