Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D9B Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB320470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320470 Supplier Novus Biologicals Supplier No. NBP298499PCP
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D9B Polyclonal antibody specifically detects TBC1D9B in Human samples. It is validated for Western Blot

Specifications

Antigen TBC1D9B
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias KIAA0676FLJ41880, MGC157757, TBC1 domain family member 9B, TBC1 domain family, member 9B (with GRAM domain)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TBC1D9B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23061
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.