Learn More
TBC1D9B Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304655AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May act as a GTPase-activating protein for Rab family protein(s).Specifications
TBC1D9B | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q66K14 | |
TBC1D9B | |
Between 1200 and 1250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TBC1D9B | |
GRAMD9B, KIAA0676, TBC1 domain family, member 9B (with GRAM domain) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23061 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.