Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBCA Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300219PE
Description
TBCA Polyclonal antibody specifically detects TBCA in Human samples. It is validated for ELISASpecifications
TBCA | |
Polyclonal | |
ELISA | |
CFA, chaperonin cofactor A, TCP1-chaperonin cofactor A, tubulin folding cofactor A, Tubulin-folding cofactor A, tubulin-specific chaperone a | |
Produced in rabbits immunized with purified, recombinant Human TBCA (Accession#: O75347; Met1-Ala108) | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
6902 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction