Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBCA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557683 Supplier Invitrogen™ Supplier No. PA557683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRAEDGENYD IKKQAEILQE SRMMIPDCQR RLEAAYLDLQ RILENEKDLE EAEEY Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBCA
Gene Accession No. O75347
Gene Alias CFA; chaperonin cofactor A; co-chaperonin associated with a- & b-tubulin; cofactor A; Tbca; Tbca13; TCP1-chaperonin cofactor A; tubulin cofactor a; tubulin folding cofactor A; tubulin-folding cofactor A; Tubulin-specific chaperone A
Gene Symbols TBCA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBCA. Recombinant protein control fragment (Product #RP-96937).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6902
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.