Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBCC Antibody (OTI4C4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274467F
Description
TBCC Monoclonal specifically detects TBCC in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TBCC | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TBCC | |
Full length recombinant protein of human TBCC (NP_003183) produced in HEK293T cells. | |
0.1 mL | |
6903.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4C4 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CFCtubulin-specific chaperone c, tubulin folding cofactor C, Tubulin-folding cofactor C | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction