Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBCD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570546
Description
This target displays homology in the following species: Human: 100%.
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications
TBCD | |
Polyclonal | |
Unconjugated | |
Tbcd | |
2310057L06Rik; A030005L14Rik; Beta-tubulin cofactor D; KIAA0988; mKIAA0988; PP1096; SSD1; SSD-1; TBCD; TFCD; tubulin folding cofactor D; tubulin-folding cofactor D; Tubulin-specific chaperone D | |
Rabbit | |
Affinity chromatography | |
RUO | |
6904 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BTW9 | |
Tbcd | |
synthetic peptide directed towards the middle region of human TBCD (aa 299-348). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction